CAS RN 537-03-1
CAS RN 537-03-1
IUPAC Name:
dilanthanum(3+) trioxalate
Synonyms:
dilanthanum(3+) trioxalate
Molecular Weight (g/mol):
541.87
Molecular Formula:
C6La2O12
InChi Key:
OXHNIMPTBAKYRS-UHFFFAOYSA-H
SMILES:
[La+3].[La+3].[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O
1
–
1
of
1
results
1
Lanthanum(III) oxalate decahydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals
CAS: 537-03-1 Molecular Formula: C6La2O12 Molecular Weight (g/mol): 541.865 MDL Number: MFCD00150540 InChI Key: OXHNIMPTBAKYRS-UHFFFAOYSA-H Synonym: lanthanum oxalate,lanthanum iii oxalate,acmc-1bafu,lanthanum 3+ ; oxalate,dilanthanum 3+ trioxalate ion,dilanthanum 3+ trioxalate,lanthanum iii oxalate decahydrate, reacton,ethanedioic acid,lanthanum 3+ salt,hydrate 3:2:9,lanthanum, m-ethanedioato 2--ko1,ko2':ko1',ko2 bis ethanedioato 2--ko1,ko2 di PubChem CID: 164654 IUPAC Name: lanthanum(3+);oxalate SMILES: C(=O)(C(=O)[O-])[O-].C(=O)(C(=O)[O-])[O-].C(=O)(C(=O)[O-])[O-].[La+3].[La+3]