accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOHOHO(E)

CAS RN 537-73-5

IUPAC Name: (2E)-3-(3-hydroxy-4-methoxyphenyl)prop-2-enoic acid
Synonyms: hesperetic acid hesperetate
Molecular Weight (g/mol): 194.19
Molecular Formula: C10H10O4
InChi Key: QURCVMIEKCOAJU-HWKANZROSA-N
SMILES: COC1=CC=C(\C=C\C(O)=O)C=C1O
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (3)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (4)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)