accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 537033-54-8

IUPAC Name: 2-(4-bromo-2,6-difluorophenyl)acetic acid
Synonyms: (4-bromo-2,6-difluorophenyl)acetic acid
Molecular Weight (g/mol): 251.03
Molecular Formula: C8H5BrF2O2
InChi Key: GBFFYYLXGIHKLU-UHFFFAOYSA-N
SMILES: OC(=O)CC1=C(F)C=C(Br)C=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results