accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 537050-14-9

IUPAC Name: 2-bromo-1-[4-fluoro-3-(trifluoromethyl)phenyl]ethan-1-one
Synonyms: 2-bromo-1-[4-fluoro-3-(trifluoromethyl)phenyl]ethanone
Molecular Weight (g/mol): 285.04
Molecular Formula: C9H5BrF4O
InChi Key: BUNOSKWLVXNWEH-UHFFFAOYSA-N
SMILES: FC1=C(C=C(C=C1)C(=O)CBr)C(F)(F)F
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results