accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOSNNHSS

CAS RN 53723-88-9

IUPAC Name: 2-[(5-sulfanylidene-4,5-dihydro-1,3,4-thiadiazol-2-yl)sulfanyl]acetate
Synonyms: 2-[(5-sulfanylidene-4H-1,3,4-thiadiazol-2-yl)sulfanyl]acetate
Molecular Weight (g/mol): 207.26
Molecular Formula: C4H3N2O2S3
InChi Key: UJBXVTJYSIDCIE-UHFFFAOYSA-M
SMILES: [O-]C(=O)CSC1=NNC(=S)S1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results