accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 53733-96-3

OOOOOONNCH3

CAS RN 53733-96-3

IUPAC Name: 2,5-dioxopyrrolidin-1-yl 2-{[(benzyloxy)carbonyl](methyl)amino}acetate
Synonyms: 2,5-dioxopyrrolidin-1-yl 2-{[(benzyloxy)carbonyl](methyl)amino}acetate
Molecular Weight (g/mol): 320.3
Molecular Formula: C15H16N2O6
InChi Key: GUHSSKCYLZIGBH-UHFFFAOYSA-N
SMILES: CN(CC(=O)ON1C(=O)CCC1=O)C(=O)OCC1=CC=CC=C1
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)