accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaNaOSOONNSOOO

CAS RN 53744-42-6

IUPAC Name: disodium 4-[7-(4-sulfonatophenyl)-1,10-phenanthrolin-4-yl]benzene-1-sulfonate
Synonyms: disodium 4-[7-(4-sulfonatophenyl)-1,10-phenanthrolin-4-yl]benzenesulfonate
Molecular Weight (g/mol): 536.48
Molecular Formula: C24H14N2Na2O6S2
InChi Key: PCNDSIWXTYFWIA-UHFFFAOYSA-L
SMILES: [Na+].[Na+].[O-]S(=O)(=O)C1=CC=C(C=C1)C1=C2C=CC3=C(C=CN=C3C2=NC=C1)C1=CC=C(C=C1)S([O-])(=O)=O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results