CAS RN 53780-34-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
PESTANAL™ Mefluidide, ≥98% (HPLC), MilliporeSigma™ Supelco™
Mefluidide is a herbicide widely used to regulate the growth of turf grass and ornamentals. Its primary use is to inhibit cell division in meristematic regions of the plant.