accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHCH3H3C

CAS RN 5379-19-1

IUPAC Name: 1-(2,4-dimethylphenyl)ethan-1-ol
Synonyms: 1-(2,4-dimethylphenyl)ethanol
Molecular Weight (g/mol): 150.22
Molecular Formula: C10H14O
InChi Key: DNHQUGRUHBFDFT-UHFFFAOYNA-N
SMILES: CC(O)C1=CC=C(C)C=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)