accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 538-24-9

IUPAC Name: 1,3-bis(dodecanoyloxy)propan-2-yl dodecanoate
Synonyms: trilaurin
Molecular Weight (g/mol): 639.02
Molecular Formula: C39H74O6
InChi Key: VMPHSYLJUKZBJJ-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCC)OC(=O)CCCCCCCCCCC
Molecular Weight (g/mol) 
  • (2)
  • (5)
Physical Form 
  • (3)
  • (4)
Quantity 
  • (1)
  • (3)
  • (2)
  • (1)
Percent Purity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (4)
  • (2)
  • (5)
  • (3)
  • (4)
  • (1)
  • (3)
  • (2)
  • (1)
  • (3)
  • (2)