accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3C

CAS RN 538-93-2

IUPAC Name: (2-methylpropyl)benzene
Synonyms: isobutylbenzene
Molecular Weight (g/mol): 134.22
Molecular Formula: C10H14
InChi Key: KXUHSQYYJYAXGZ-UHFFFAOYSA-N
SMILES: CC(C)CC1=CC=CC=C1
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)