accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClPOOO

CAS RN 5381-99-7

IUPAC Name: 2-chloro-2,4-dihydro-1,3,2-benzodioxaphosphinin-4-one
Synonyms: 2-chloro-1,3,2-benzodioxaphosphinin-4-one
Molecular Weight (g/mol): 202.53
Molecular Formula: C7H4ClO3P
InChi Key: BVOITXUNGDUXRW-UHFFFAOYNA-N
SMILES: ClP1OC(=O)C2=CC=CC=C2O1
Molecular Weight (g/mol) 
  • (4)
  • (2)
Quantity 
  • (3)
  • (3)
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)