accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5382-17-2

ClOHNH2

CAS RN 5382-17-2

IUPAC Name: 4-hydroxypiperidin-1-ium chloride
Synonyms: 4-hydroxypiperidin-1-ium chloride
Molecular Weight (g/mol): 137.61
Molecular Formula: C5H12ClNO
InChi Key: VKCORPXOKYDINR-UHFFFAOYSA-N
SMILES: [Cl-].OC1CC[NH2+]CC1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)