accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5382-23-0

IUPAC Name: hydrogen 4-chloro-1-methylpiperidine chloride
Synonyms: hydrogen 4-chloro-1-methylpiperidine chloride
Molecular Weight (g/mol): 170.08
Molecular Formula: C6H13Cl2N
InChi Key: QDOJNWFOCOWAPO-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CN1CCC(Cl)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results