accessibility menu, dialog, popup

UserName

Viewing profile as user:
NHOOHCl

CAS RN 53843-90-6

IUPAC Name: benzyl pyrrolidine-2-carboxylate hydrochloride
Synonyms: benzyl pyrrolidine-2-carboxylate hydrochloride
Molecular Weight (g/mol): 241.72
Molecular Formula: C12H16ClNO2
InChi Key: NEDMOHHWRPHBAL-UHFFFAOYNA-N
SMILES: Cl.O=C(OCC1=CC=CC=C1)C1CCCN1
Grade 
  • (2)
  • (1)
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)