accessibility menu, dialog, popup

UserName

Viewing profile as user:
SOOBr

CAS RN 53846-85-8

IUPAC Name: 12-bromo-8λ⁶-thiatricyclo[7.4.0.0²,⁷]trideca-1(9),2,4,6,10,12-hexaene-8,8-dione
Synonyms: 12-bromo-8λ⁶-thiatricyclo[7.4.0.0²,⁷]trideca-1(9),2,4,6,10,12-hexaene-8,8-dione
Molecular Weight (g/mol): 295.15
Molecular Formula: C12H7BrO2S
InChi Key: WRKJLRFOQLWYOW-UHFFFAOYSA-N
SMILES: BrC1=CC2=C(C=C1)S(=O)(=O)C1=CC=CC=C21
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results