accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 53857-57-1

IUPAC Name: 5-bromo-1H-indazole
Synonyms: 5-bromo-1H-indazole
Molecular Weight (g/mol): 197.04
Molecular Formula: C7H5BrN2
InChi Key: STVHMYNPQCLUNJ-UHFFFAOYSA-N
SMILES: BrC1=CC=C2NN=CC2=C1
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (4)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)