accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 53874-66-1

IUPAC Name: 1-(chloromethyl)-3-phenoxybenzene
Synonyms: 3-phenoxybenzyl chloride
Molecular Weight (g/mol): 218.68
Molecular Formula: C13H11ClO
InChi Key: QUYVTGFWFHQVRO-UHFFFAOYSA-N
SMILES: ClCC1=CC=CC(OC2=CC=CC=C2)=C1
Boiling Point 
  • (3)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)