accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 53896-49-4

NNN

CAS RN 53896-49-4

IUPAC Name: pyridazine-3-carbonitrile
Synonyms: pyridazine-3-carbonitrile
Molecular Weight (g/mol): 105.1
Molecular Formula: C5H3N3
InChi Key: PJESVVYWPFAJCS-UHFFFAOYSA-N
SMILES: N#CC1=NN=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results