CAS RN 539-90-2
IUPAC Name:
2-methylpropyl butanoate
Synonyms:
isobutyl butyrate
Molecular Weight (g/mol):
144.21
Molecular Formula:
C8H16O2
InChi Key:
RGFNRWTWDWVHDD-UHFFFAOYSA-N
SMILES:
CCCC(=O)OCC(C)C