accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 53911-92-5

IUPAC Name: tributyl(3-methylbut-2-en-1-yl)stannane
Synonyms: tributyl(3-methylbut-2-en-1-yl)stannane
Molecular Weight (g/mol): 359.19
Molecular Formula: C17H36Sn
InChi Key: XEFRYQPTIWSVGI-UHFFFAOYSA-N
SMILES: CCCC[Sn](CCCC)(CCCC)CC=C(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)