accessibility menu, dialog, popup

UserName

Viewing profile as user:
NHO

CAS RN 53912-80-4

IUPAC Name: (1-benzylpyrrolidin-2-yl)methanol
Synonyms: (1-benzylpyrrolidin-2-yl)methanol
Molecular Weight (g/mol): 191.27
Molecular Formula: C12H17NO
InChi Key: ZAIQBJPTOXDDKA-UHFFFAOYNA-N
SMILES: OCC1CCCN1CC1=CC=CC=C1
Boiling Point 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)