accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClOHO(S)HNH

CAS RN 53912-85-9

IUPAC Name: hydrogen 2-[(2S)-pyrrolidin-2-yl]acetic acid chloride
Synonyms: hydrogen (2S)-pyrrolidin-2-ylacetic acid chloride
Molecular Weight (g/mol): 165.62
Molecular Formula: C6H12ClNO2
InChi Key: VQDACVOAOJQTPR-JEDNCBNOSA-N
SMILES: [H+].[Cl-].OC(=O)C[C@@H]1CCCN1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)