accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClClNCO

CAS RN 5392-82-5

IUPAC Name: 1,4-dichloro-2-isocyanatobenzene
Synonyms: 1,4-dichloro-2-isocyanatobenzene
Molecular Weight (g/mol): 188.01
Molecular Formula: C7H3Cl2NO
InChi Key: PEQMJVGRHNZPAM-UHFFFAOYSA-N
SMILES: ClC1=CC=C(Cl)C(=C1)N=C=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results