accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5396-18-9

IUPAC Name: 1-(2-hydroxy-3,4-dimethoxyphenyl)ethan-1-one
Synonyms: 1-(2-hydroxy-3,4-dimethoxyphenyl)ethanone
Molecular Weight (g/mol): 196.2
Molecular Formula: C10H12O4
InChi Key: BCEPNLMYVYJIHU-UHFFFAOYSA-N
SMILES: COC1=CC=C(C(C)=O)C(O)=C1OC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results