accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 53978-73-7

IUPAC Name: (2R)-2-{[(benzyloxy)carbonyl](methyl)amino}-3-methylbutanoic acid
Synonyms: (2R)-2-{[(benzyloxy)carbonyl](methyl)amino}-3-methylbutanoic acid|Z-D-N-ME-VAL-OH
Molecular Weight (g/mol): 265.31
Molecular Formula: C14H19NO4
InChi Key: NNEHOKZDWLJKHP-GFCCVEGCSA-N
SMILES: CC(C)[C@@H](N(C)C(=O)OCC1=CC=CC=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)