accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOClNCl

CAS RN 5398-44-7

IUPAC Name: 2,6-dichloropyridine-4-carboxylic acid
Synonyms: 2,6-dichloropyridine-4-carboxylic acid
Molecular Weight (g/mol): 192
Molecular Formula: C6H3Cl2NO2
InChi Key: SQSYNRCXIZHKAI-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(Cl)=NC(Cl)=C1
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)