accessibility menu, dialog, popup

UserName

Viewing profile as user:
FHOH2N

CAS RN 53981-25-2

IUPAC Name: 2-amino-6-fluorophenol
Synonyms: 2-amino-6-fluorophenol
Molecular Weight (g/mol): 127.12
Molecular Formula: C6H6FNO
InChi Key: MDIAVSZFIQWYST-UHFFFAOYSA-N
SMILES: NC1=C(O)C(F)=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results