accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 53994-85-7

ClOOOHONHCH3CH3H3C(R)

CAS RN 53994-85-7

IUPAC Name: (2R)-2-{[(tert-butoxy)carbonyl]amino}-2-(4-chlorophenyl)acetic acid
Synonyms: (R)-[(tert-butoxycarbonyl)amino](4-chlorophenyl)acetic acid
Molecular Weight (g/mol): 285.72
Molecular Formula: C13H16ClNO4
InChi Key: ZZONJNNLTAGSHB-SNVBAGLBSA-N
SMILES: CC(C)(C)OC(=O)N[C@@H](C(O)=O)C1=CC=C(Cl)C=C1
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)