CAS RN 54-06-8
CAS RN 54-06-8
IUPAC Name:
3-hydroxy-1-methyl-2,3,5,6-tetrahydro-1H-indole-5,6-dione
Synonyms:
adrenochrome
Molecular Weight (g/mol):
179.18
Molecular Formula:
C9H9NO3
InChi Key:
RPHLQSHHTJORHI-UHFFFAOYNA-N
SMILES:
CN1CC(O)C2=CC(=O)C(=O)C=C12
1
–
1
of
1
results
1
Thermo Scientific Chemicals 3-Hydroxy-1-methyl-5,6-indolinedione, 98%
CAS: 54-06-8 Molecular Formula: C9H9NO3 Molecular Weight (g/mol): 179.17 MDL Number: MFCD00069732 InChI Key: RPHLQSHHTJORHI-UHFFFAOYSA-N Synonym: adrenochrome,adraxone,usaf uctl-7,3-hydroxy-1-methyl-5,6-indolinedione,1-adrenochrome,3-hydroxy-1-methylindoline-5,6-dione,2,3-dihydro-3-hydroxy-1-methyl-1h-indole-5,6-dione,5,6-indolinedione, 3-hydroxy-1-methyl,adrenochrome, l,adrenochrom PubChem CID: 5898 IUPAC Name: 3-hydroxy-1-methyl-2,3-dihydroindole-5,6-dione SMILES: CN1CC(C2=CC(=O)C(=O)C=C21)O