accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3N(S)N

CAS RN 54-11-5

IUPAC Name: 3-[(2S)-1-methylpyrrolidin-2-yl]pyridine
Synonyms: nicoderm CQ bitartrate, nicotine nicotine agent micotine caswell no. 597 tetrahydronicotyrine, dl- methyl-2-pyrrolidinyl)pyridine XL all insecticide rcra waste number P075 nicotine agent (substance)
Molecular Weight (g/mol): 162.24
Molecular Formula: C10H14N2
InChi Key: SNICXCGAKADSCV-JTQLQIEISA-N
SMILES: CN1CCC[C@H]1C1=CC=CN=C1
Molecular Weight (g/mol) 
  • (4)
  • (7)
Physical Form 
  • (10)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (4)
  • (3)
Percent Purity 
  • (3)
  • (2)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (1)
  • (1)
  • (3)
  • (6)
  • (1)
  • (4)
  • (7)
  • (10)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)
  • (3)
  • (3)
  • (2)
  • (4)
  • (1)
  • (2)
  • (1)
  • (1)