accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 54-42-2

IUPAC Name: 1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-iodo-1,2,3,4-tetrahydropyrimidine-2,4-dione
Synonyms: idoxuridine|5-iododeoxyuridine|IdU|antizona|herplex|stoxil|emanil|herpid|idoxuridin|dendrid
Molecular Weight (g/mol): 354.1
Molecular Formula: C9H11IN2O5
InChi Key: XQFRJNBWHJMXHO-RRKCRQDMSA-N
SMILES: OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(I)C(=O)NC1=O
Quantity 
  • (4)
  • (3)
  • (1)
  • (4)
Physical Form 
  • (6)
  • (4)
Percent Purity 
  • (3)
  • (1)
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (4)
  • (3)
  • (5)
  • (4)
  • (3)
  • (1)
  • (4)
  • (6)
  • (4)
  • (3)
  • (1)
  • (2)
  • (3)