accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 54-62-6

IUPAC Name: 2-[(4-{[(2,4-diaminopteridin-6-yl)methyl]amino}phenyl)formamido]pentanedioic acid
Synonyms: aminopterin 氨蝶呤钠 aminopterin sodium
Molecular Weight (g/mol): 440.42
Molecular Formula: C19H20N8O5
InChi Key: TVZGACDUOSZQKY-UHFFFAOYNA-N
SMILES: NC1=NC(N)=C2N=C(CNC3=CC=C(C=C3)C(=O)NC(CCC(O)=O)C(O)=O)C=NC2=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results