CAS RN 54-77-3
IUPAC Name:
1,1-dimethyl-4-phenylpiperazin-1-ium iodide
Synonyms:
DMPP dimethylphenylpiperazinium iodide dmpp iodide
Molecular Weight (g/mol):
318.2
Molecular Formula:
C12H19IN2
InChi Key:
XFZJGFIKQCCLGK-UHFFFAOYSA-M
SMILES:
[I-].C[N+]1(C)CCN(CC1)C1=CC=CC=C1
1
–
1
of
1
results
1
1,1-Dimethyl-4-phenylpiperazinium Iodide 99%, Thermo Scientific™
CAS: 54-77-3 Molecular Formula: C12H19IN2 Molecular Weight (g/mol): 318.20 MDL Number: MFCD00011976 InChI Key: XFZJGFIKQCCLGK-UHFFFAOYSA-M Synonym: 1,1-dimethyl-4-phenylpiperazinium iodide,dmpp,dmpp iodide,dimethylphenylpiperazinium iodide,1,1-dimethyl-4-phenylpiperazin-1-ium iodide,1,1-dimethyl-4-phenylpiperazine iodide,n,n-dimethyl-n'-phenylpiperazinium iodide,dimethyl-4-phenylpiperazium iodide,1,1-dimethyl-4-phenyl-piperazinium iodide,unii-x95h7drx99 PubChem CID: 5911 ChEBI: CHEBI:4290 IUPAC Name: 1,1-dimethyl-4-phenylpiperazin-1-ium iodide SMILES: [I-].C[N+]1(C)CCN(CC1)C1=CC=CC=C1