accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2NH2N

CAS RN 54-96-6

IUPAC Name: pyridine-3,4-diamine
Synonyms: 3,4-diaminopyridine amifampridine
Molecular Weight (g/mol): 109.13
Molecular Formula: C5H7N3
InChi Key: OYTKINVCDFNREN-UHFFFAOYSA-N
SMILES: NC1=CC=NC=C1N
Molecular Weight (g/mol) 
  • (4)
  • (5)
Percent Purity 
  • (5)
  • (2)
  • (2)
Quantity 
  • (3)
  • (2)
  • (4)
Physical Form 
  • (4)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (2)
  • (5)
  • (4)
  • (5)
  • (5)
  • (2)
  • (2)
  • (3)
  • (2)
  • (4)
  • (4)
  • (1)