accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 540-10-3

IUPAC Name: hexadecyl hexadecanoate
Synonyms: cetyl palmitate
Molecular Weight (g/mol): 480.86
Molecular Formula: C32H64O2
InChi Key: PXDJXZJSCPSGGI-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCOC(=O)CCCCCCCCCCCCCCC
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
Boiling Point 
  • (3)
  • (2)
Physical Form 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (5)
  • (1)