accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClCH3NH2

CAS RN 540-23-8

IUPAC Name: hydrogen 4-methylaniline chloride
Synonyms: toluidinium chloride
Molecular Weight (g/mol): 143.61
Molecular Formula: C7H10ClN
InChi Key: JQKBUTDZZRGQDR-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CC1=CC=C(N)C=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)