accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH3HOO

CAS RN 540-69-2

IUPAC Name: formic acid amine
Synonyms: ammonium formate
Molecular Weight (g/mol): 63.06
Molecular Formula: CH5NO2
InChi Key: VZTDIZULWFCMLS-UHFFFAOYSA-N
SMILES: N.OC=O
Molecular Weight (g/mol) 
  • (1)
  • (11)
  • (12)
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)
  • (2)
  • (4)
  • (3)
  • (1)
Percent Purity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (11)
  • (2)
  • (3)
Physical Form 
  • (4)
  • (1)
  • (6)
  • (5)
  • (1)
  • (2)
Grade 
  • (1)
  • (2)
  • (1)
  • (5)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (5)
  • (3)
  • (4)
  • (1)
  • (5)
  • (11)
  • (1)
  • (1)
  • (11)
  • (12)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)
  • (2)
  • (4)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (11)
  • (2)
  • (3)
  • (4)
  • (1)
  • (6)
  • (5)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (5)
  • (1)