accessibility menu, dialog, popup

UserName

Viewing profile as user:
ICH3NH3CCH3

CAS RN 5400-94-2

IUPAC Name: benzyltriethylazanium iodide
Synonyms: benzyltriethylazanium iodide
Molecular Weight (g/mol): 319.23
Molecular Formula: C13H22IN
InChi Key: JWLJBISFJGEYMT-UHFFFAOYSA-M
SMILES: [I-].CC[N+](CC)(CC)CC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results