accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 54013-04-6

IUPAC Name: ethyl 5-chloropyrazine-2-carboxylate
Synonyms: ethyl 5-chloropyrazine-2-carboxylate
Molecular Weight (g/mol): 186.6
Molecular Formula: C7H7ClN2O2
InChi Key: CFKGDFQQELWOHA-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CN=C(Cl)C=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results