accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5402-73-3

ClClSClOO

CAS RN 5402-73-3

IUPAC Name: 2,5-dichlorobenzene-1-sulfonyl chloride
Synonyms: 2,5-dichlorobenzenesulfonyl chloride
Molecular Weight (g/mol): 245.5
Molecular Formula: C6H3Cl3O2S
InChi Key: BXCOSWRSIISQSL-UHFFFAOYSA-N
SMILES: ClC1=CC=C(Cl)C(=C1)S(Cl)(=O)=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)