accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 54039-38-2

IUPAC Name: dichlorozirconium; bis(pentamethylcyclopenta-2,4-dien-1-ide)
Synonyms: dichlorozirconium; bis(pentamethylcyclopenta-2,4-dien-1-ide)
Molecular Weight (g/mol): 432.59
Molecular Formula: C20H30Cl2Zr
InChi Key: OCFSLQKTBFSWPL-UHFFFAOYSA-L
SMILES: Cl[Zr]Cl.C[C-]1C(C)=C(C)C(C)=C1C.C[C-]1C(C)=C(C)C(C)=C1C
Percent Purity 
  • (1)
  • (1)
Physical Form 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)