accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 540792-53-8

IUPAC Name: N-(2,4-dibromophenyl)-2-methoxybenzamide
Synonyms: N-(2,4-dibromophenyl)-2-methoxybenzamide
Molecular Weight (g/mol): 385.06
Molecular Formula: C14H11Br2NO2
InChi Key: MXCGGFUISNEJRG-UHFFFAOYSA-N
SMILES: COC1=CC=CC=C1C(=O)NC1=C(Br)C=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results