accessibility menu, dialog, popup

UserName

Viewing profile as user:
NClO

CAS RN 54087-03-5

IUPAC Name: 6-chloropyridine-2-carbaldehyde
Synonyms: 6-chloropyridine-2-carbaldehyde
Molecular Weight (g/mol): 141.55
Molecular Formula: C6H4ClNO
InChi Key: XTRLIKXVRGWTKW-UHFFFAOYSA-N
SMILES: ClC1=CC=CC(C=O)=N1
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)