accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5409-60-9

H3CO

CAS RN 5409-60-9

IUPAC Name: 4,4-diphenylbutan-2-one
Synonyms: 4,4-diphenylbutan-2-one
Molecular Weight (g/mol): 224.3
Molecular Formula: C16H16O
InChi Key: FPHXYKLKNOEKTQ-UHFFFAOYSA-N
SMILES: CC(=O)CC(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results