accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFSOOClCl

CAS RN 54090-08-3

IUPAC Name: 2-chloro-5-(trifluoromethyl)benzene-1-sulfonyl chloride
Synonyms: 2-chloro-5-(trifluoromethyl)benzenesulfonyl chloride
Molecular Weight (g/mol): 279.05
Molecular Formula: C7H3Cl2F3O2S
InChi Key: ZEYKLMDPUOVUCR-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(=C(Cl)C=C1)S(Cl)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results