accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OPOOOFFFOCH3

CAS RN 54094-19-8

IUPAC Name: dimethyl {2-oxo-3-[3-(trifluoromethyl)phenoxy]propyl}phosphonate
Synonyms: dimethyl 2-oxo-3-[3-(trifluoromethyl)phenoxy]propylphosphonate
Molecular Weight (g/mol): 326.21
Molecular Formula: C12H14F3O5P
InChi Key: MBGWNNJXMYHKQO-UHFFFAOYSA-N
SMILES: COP(=O)(CC(=O)COC1=CC=CC(=C1)C(F)(F)F)OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results