accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SiH3COSiCH3CH3OSiCH3CH3OSiCH3H3COSiH3CCH3O

CAS RN 541-02-6

IUPAC Name: decamethyl-1,3,5,7,9,2,4,6,8,10-pentaoxapentasilecane
Synonyms: decamethylcyclopentasiloxane
Molecular Weight (g/mol): 370.77
Molecular Formula: C10H30O5Si5
InChi Key: XMSXQFUHVRWGNA-UHFFFAOYSA-N
SMILES: C[Si]1(C)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)O1
Molecular Weight (g/mol) 
  • (9)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (1)
  • (6)
  • (2)
Percent Purity 
  • (3)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (4)
  • (9)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (6)
  • (2)
  • (3)
  • (4)
  • (6)
  • (1)
  • (1)