accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 541-22-0

IUPAC Name: trimethyl[10-(trimethylazaniumyl)decyl]azanium dibromide
Synonyms: decamethonium bromide
Molecular Weight (g/mol): 418.3
Molecular Formula: C16H38Br2N2
InChi Key: HLXQFVXURMXRPU-UHFFFAOYSA-L
SMILES: [Br-].[Br-].C[N+](C)(C)CCCCCCCCCC[N+](C)(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results