CAS RN 541-31-1
IUPAC Name:
3-methylbutane-1-thiol
Synonyms:
isoamylthiol
Molecular Weight (g/mol):
104.21
Molecular Formula:
C5H12S
InChi Key:
GIJGXNFNUUFEGH-UHFFFAOYSA-N
SMILES:
CC(C)CCS
1
–
1
of
1
results
1
Isoamyl Mercaptan 94.0+%, TCI America™
CAS: 541-31-1 Molecular Formula: C5H12S Molecular Weight (g/mol): 104.211 MDL Number: MFCD00004899 InChI Key: GIJGXNFNUUFEGH-UHFFFAOYSA-N Synonym: isoamyl mercaptan,3-methyl-1-butanethiol,1-butanethiol, 3-methyl,isopentyl mercaptan,3-methylbutanethiol,isoamylthiol,isopentanethiol,isoamylmercaptan,unii-mmk4sun45e,isoamyl sulfhydrate PubChem CID: 10925 IUPAC Name: 3-methylbutane-1-thiol SMILES: CC(C)CCS